Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PPP2R5B Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA112390

Catalog No. 01-674-123


Add to cart

Description

Description

Protein phosphatase 2A is one of the four major Ser/Thr phosphatases and is implicated in the negative control of cell growth and division. Protein phosphatase 2A holoenzymes are heterotrimeric proteins composed of a structural subunit A, a catalytic subunit C, and a regulatory subunit B. The regulatory subunit is encoded by a diverse set of genes that have been grouped into the B/PR55, B ft./PR61, and B ft. ft./PR72 families. These different regulatory subunits confer distinct enzymatic specificities and intracellular localizations to the holozenzyme. The product of this gene belongs to the B ft. family. This gene encodes a specific phosphotyrosyl phosphatase activator of the dimeric form of protein phosphatase 2A. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications

Specifications

PPP2R5B
Polyclonal
PBS with 0.08% sodium azide
Q15173, Q6PD28, Q80W83
Serine/threonine-protein phosphatase 2A regulatory subunit B′′ subunit beta, PP2A subunit B isoform PR48, Protein phosphatase 2A 48 kDa regulatory subunit, Protein Phosphatase 2 A/B (B′ pan 2)
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
225849, 309179, 505915, 5526
Western Blot
Unconjugated
PPP2R5B
Liquid
PPP2R5B
Synthetic peptide.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit