Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PP2A alpha, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA133120

Catalog No. PIPA133120

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-33120 detects Protein phosphatase 2 from human samples.PA1-33120 has been successfully used in Western blot applications.The PA1-33120 immunogen is: Synthetic peptide EPHVTRRTPDYFL derived from C terminal sequence of human protein.
Specifications

Specifications

Protein phosphatase 2
Polyclonal
Unconjugated
PPP2CA
Purified
Goat
IgG
100μg
Store at 4°C or Store at -20°C if preferred
Primary
5515
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
P67775, P63331
PPP2CA
Synthetic peptide EPHVTRRTPDYFL derived from C terminal sequence of human protein.
IgG fraction
RUO
Antibody
Polyclonal
Canine, Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit