Learn More
Prothrombin Sheep anti-Human, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA143039
Catalog No. PA143039
Add to cart
Description
Description
PA1-43039 detects Prothrombin in human samples. PA1-43039 has successfully been used in Western blot and ELISA applications. PA1-43039 was derived from human prothrombin.
Prothrombin or Coagulation factor II, is proteolytically cleaved to form thrombin in the first step of the coagulation cascade which ultimately results in the stemming of blood loss. F2 also plays a role in maintaining vascular integrity during development and postnatal life. Mutations in F2 leads to various forms of thrombosis and dysprothrombinemia.
Specifications
Specifications
Prothrombin
Polyclonal
Unconjugated
F2
Liquid
F2
Human Prothrombin.
Ammonium sulfate precipitation, Ion-exchange chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Western Blot
8.2 mg/mL
50%water with 50% glycerol and no preservative
P00734
F2
Sheep
IgG
1 mg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2147