Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Protocadherin-15, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner 46010002

Catalog No. 46-010-002

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protocadherin-15 Polyclonal antibody specifically detects Protocadherin-15 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Protocadherin-15
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
PCDH15
This antibody is specific for the C Terminus Region of the target protein.
Immunogen affinity purified
RUO
Primary
65217
Human
Western Blot, ELISA
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
Q96QU1
autosomal recessive 23, cadherin-related family member 15, DKFZp667A1711, protocadherin-15, protocadherin-related 15
Rabbit
IgG
0.05 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human PCDH15.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only