anti-Protocadherin-15, Polyclonal, Novus Biologicals™

Manufacturer: Novus BiologicalsSupplier Diversity Partner 209500020.1MG

 View more versions of this product

Catalog No. 20-950-00201

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Host Species Rabbit
    Applications ELISA,Western Blotting
    Quantity 0.1mg
    Regulatory Status RUO
    Cross Reactivity Human
    Antigen Protocadherin-15
    Format Affinity Purified
    Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0
    Gene Accession No. Q96QU1
    Immunogen This antibody is specific for the N Terminus Region of the target protein (NM_023115).
    Monoclonal or Polyclonal Polyclonal
    Gene ID (Entrez) 65217
    Test Specificity This product is specific for Human PCDH15.
    Conjugate Unlabeled
    Primary or Secondary Primary

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit