Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
PCDHGA9 Mouse anti-Human, Clone: 1G10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA522248
Catalog No. PIMA522248
Description
Description
Peptide Sequence: VREDAPQGTV ILLFNAHDRD SGKNGQVVCS IQENLSFTLE NSEEDYYRLL TAQILDREKA SEYNITVTAT DRGTPPLSTE IHITLQVTDI
This gene is a member of the protocadherin gamma gene cluster, one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes. The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes.
Specifications
Specifications
PCDHGA9
Monoclonal
0.2-1.0 mg/mL
PBS with no preservative; pH 7.4
Q9Y5G4
PAK1, PKN, PRK1, PRKCL1
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
56107
ELISA, Western Blot
1G10
Unconjugated
PCDHGA9
Liquid
PCDHGA9
PCDHGA9 (NP_061744, 356 a.a. ∽ 445 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human