Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PROX1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511898

Catalog No. PIPA511898

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken based on sequence homology.

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections.
Specifications

Specifications

PROX1
Polyclonal
PBS with 0.09% sodium azide
Q92786, P48437
PROX1
KLH conjugated synthetic peptide between 144-175 amino acids from the N-terminal region of human PROX1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
Unconjugated
PROX1
Liquid
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
19130, 5629
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit