Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRP19 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524797

Catalog No. PIPA524797

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine, chicken and rat based on sequence homology.

PRPF19 is the human homolog of yeast Pso4, a gene essential for cell survival and DNA repair.
Specifications

Specifications

PRP19
Polyclonal
PBS with 0.09% sodium azide
Q9UMS4, Q99KP6
NMP200, PRP19, PSO4, SNEV, UBOX4, hPSO4, PRP19/PSO4 homolog, nuclear matrix protein 200, nuclear matrix protein NMP200 related to splicing factor PRP19, pre-mRNA-processing factor 19, senescence evasion factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27339, 28000
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
PRPF19
Liquid
PRPF19
KLH conjugated synthetic peptide between 4-33 amino acids from the N-terminal region of human PRPF19
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit