Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527749

Catalog No. PIPA527749


Add to cart

Description

Description

PA5-27749 targets PRPS1 in WB applications and shows reactivity with Human samples. The PA5-27749 immunogen is a recombit protein fragment corresponding to a region within amino acids 1 and 318 of PRPS1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome.
Specifications

Specifications

PRPS1
Polyclonal
Unconjugated
PRPS1
Liquid
PRPS1
Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 318 of PRPS1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry (Paraffin), Western Blot
0.58 mg/mL
tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
P60891, Q9D7G0, P60892
ARTS, CMTX5, DFN2, DFNX1, PPRibP, PRS-I, PRSI, dJ1070B1.2 (phosphoribosyl pyrophosphate synthetase 1), deafness 2, perceptive, congenital, deafness, X-linked 2, perceptive, congenital, phosphoribosyl pyrophosphate synthase I, ribose-phosphate diphosphokinase 1, ribose-phosphate pyrophosphokinase 1
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
19139, 29562, 5631
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit