Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PRR5L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521129
Description
A suggested positive control is NIH-3T3 cell lysate. PA5-21129 can be used with blocking peptide PEP-1243.
PRR5L Polyclonal antibody specifically detects PRR5L in Human, Mouse samples. It is validated for Immunocytochemistry, Western Blot.Specifications
PRR5L | |
Polyclonal | |
Unconjugated | |
PRR5L | |
2600010E01Rik; 4833411O04Rik; proline rich 5 like; proline-rich protein 5-like; Protein observed with Rictor-2; PROTOR2; protor-2; Prr5l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
72446, 79899 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
A2AVJ5, Q6MZQ0 | |
PRR5L | |
A 17 amino acid peptide near the center of human PRR5L. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction