Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRRG3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524122

Catalog No. PIPA524122

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PRRG3
Polyclonal
PBS with 0.09% sodium azide
Q9BZD7
PRGP3, TMG3, proline-rich Gla protein 3, proline-rich gamma-carboxyglutamic acid protein 3, transmembrane gamma-carboxyglutamic acid protein 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79057
Flow Cytometry, Western Blot
Unconjugated
PRRG3
Liquid
PRRG3
KLH conjugated synthetic peptide between 121-150 amino acids from the central region of human PRRG3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit