Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trypsin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524228

Catalog No. PIPA524228


Add to cart

Description

Description

This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is secreted by the pancreas and cleaved to its active form in the small intestine. It is active on peptide linkages involving the carboxyl group of lysine or arginine. Mutations in this gene are associated with hereditary pancreatitis. This gene and several other trypsinogen genes are localized to the T cell receptor beta locus on chromosome 7.
Specifications

Specifications

Trypsin
Polyclonal
PBS with 0.09% sodium azide
P07477
TRP1, TRY1, TRY4, TRYP1, beta-trypsin, cationic trypsinogen, digestive zymogen, nonfunctional trypsin 1, trypsin-1, trypsinogen 1, trypsinogen A
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5644
Western Blot
Unconjugated
PRSS1
Liquid
PRSS1
KLH conjugated synthetic peptide between 81-107 amino acids from the central region of human PRSS1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit