Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PSAP, Polyclonal, R&D Systems
Rabbit Polyclonal Antibody
Manufacturer: R&D Systems AF8520
This item is not returnable.
View return policy
Description
PSAP Polyclonal antibody specifically detects PSAP in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
PSAP | |
Unconjugated | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. See Certificate of Analysis for details. with Sodium Azide | |
Purified | |
PSAP | |
S. frugiperda insect ovarian cell line Sf21-derived recombinant human PSAP Gly17-Asn524 Accession # P07602 | |
Immunogen affinity purified | |
RUO | |
Primary | |
5660 | |
Human |
Immunocytochemistry, Western Blot | |
Western Blot 1:1000 dilution, Immunocytochemistry 1:100 dilution | |
P07602 | |
FLJ00245, GLBA, proactivator polypeptide, prosaposin, SAP1, SAP1MGC110993, sphingolipid activator protein-1 | |
Rabbit | |
IgG | |
100 uL | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC, as supplied. 1 month, 2 to 8 degreesC under sterile conditions after opening. 6 months, -20 to -70 degreesC under sterile conditions after opening. | |
Polyclonal | |
Detects human PSAP in direct ELISAs and Western blots. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only