Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PSCA, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP128492

Catalog No. NBP128492


Add to cart

Description

Description

PSCA Polyclonal antibody specifically detects PSCA in Human samples. It is validated for Peptide ELISA.
Specifications

Specifications

PSCA
0.5 mg/mL
Peptide ELISA Detection limit 1:1000
NP_005663.1
PRO232, prostate stem cell antigen
Goat
IgG
0.1 mg
Cancer, Prostate Cancer
Primary
8000
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
PSCA
Peptide with sequence KAQVSNEDCLQVEN corresponding to internal region according to NP_005663.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only