Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PSD95, Clone: 6G6-1C9, Abcam™ Non-distribution product as customer accommodation.

Manufacturer:  Abcam AB2723100UG

Catalog No. NC0122882


Add to cart

Description

Description

Host Species: Mouse
Clone: 6G6-1C9
Isotype: IgG2a
Cross Reactivity: Mouse, Rat, Human Predicted to work with Cow
Immunogen: Purified recombinant rat PSD-95

Applications:

Flow Cytometry, Western Blotting

Specifications

Specifications

PSD95
1mg/mL
Murine
IgG2a
Neuroscience
Monoclonal
6G6-1C9
Unconjugated
Purified recombinant rat PSD-95
100μg
Primary
Bovine, Human, Murine, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit