Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pseudomonas Rabbit anti-Bacteria, HRP, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA173118

Catalog No. PA173118


Add to cart

Description

Description

The PA1-73118 antibody reacts with Pseudomonas species. PA1-73118 has been successfully used in ELISA applications. The PA1-73118 antibody was raised against whole cells of Pseudomonas aeruginosa, ATCC No.27853.

Specifications

Specifications

Pseudomonas
Polyclonal
HRP
Liquid
Whole cells of Pseudomonas aeruginosa, ATCC No.27853.
Purified
RUO
Antibody
Polyclonal
ELISA
1-2 mg/mL
PBS with 10mg/mL BSA and 0.002% thimerosal
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit