Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PSG4, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP145396

Catalog No. NBP145396

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

PSG4
Unconjugated
PBS (pH 7.2), 0.5% BSA and 30% Glycerol with 0.01% Thimerosal
Affinity Purified
PSG4
Recombinant human BMP-2.
Immunogen affinity purified
RUO
Primary
5672
Western Blot
Western Blot
EAX10387
pregnancy specific beta-1-glycoprotein 4, pregnancy-specific beta-1-glycoprotein 4, PSBG-4
Rabbit
IgG
0.1 mg
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only