Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PSMC3IP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP192301

Catalog No. NBP192301


Add to cart

Description

Description

PSMC3IP Polyclonal antibody specifically detects PSMC3IP in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

PSMC3IP
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
DBD-interacting, GT198, GT198 alternative, homologous-pairing protein 2 homolog, HOP2, HUMGT198A, Nuclear receptor coactivator GT198, Proteasome 26S ATPase subunit 3-interacting protein, PSMC3 interacting protein, Tat-binding protein 1-interacting protein, TBP-1 interacting protein, TBP-1-interacting protein, TBPIPPSMC3-interacting protein
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Specificity of human PSMC3IP antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 0.4 ug/ml, Simple Western 1:20, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500
Affinity Purified
PSMC3IP
This antibody was developed against Recombinant Protein corresponding to amino acids:PEEKEQVYRERQKYCKEWRKRKRMATELSDAILEGYPKSKKQFFEEVGIETDEDYNVTLPD
Immunogen affinity purified
RUO
Primary
29893
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only