Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMC5, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127903

Catalog No. PIPA127903

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The 26S protease is involved in the ATP-dependent degradation of ubiquitinated proteins. The regulatory (or ATPase) complex confers ATP dependency and substrate specificity to the 26S complex. The 26S proteasome is a multicatalytic proteinase complex with

PA1-27903 detects PSMC5 from human, mouse, rat, bovine, and porcine samples.PA1-27903 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27755 immunogen is a synthetic peptide corresponding to human 26S protease regulatory subunit 8.
Specifications

Specifications

PSMC5
Polyclonal
PBS with no preservative
P62194, P62195, P62196, P62197, P62198
S8, SUG-1, SUG1, TBP10, TRIP1, p45, p45/SUG
Chicken
IgY
50μg
Store at -20°C
Primary
5705
Western Blot
Unconjugated
PSMC5
Purified
PSMC5
Synthetic peptide corresponding to human 26S protease regulatory subunit 8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Human, Murine, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit