Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

proteasome (prosome, macropain) 26S subunit, non-ATPase, 8 (B02), Mouse anti-Human, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PSMD8 protein.

Manufacturer:  Abnova Corporation H00005714B02

Catalog No. 89-006-072


Add to cart

Description

Description

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 nonidentical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a nonlysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. A pseudogene has been identified on chromosome 1. [provided by RefSeq

Sequence: MYEQLKGEWNRKSPNLSKCGEELGRLKLVLLELNFLPTTGTKLTKQQLILARDILEIGAQWSILRKDIPSFERYMAQLKCYYFDYKEQLPESAYMHQLLGLNLLFLLSQNRVAEFHTELERLPAKDIQTNVYIKHPVSLEQYLMEGSYNKVFLAKGNIPAESYTFFIDILLDTIRDEIAGCIEKAYEKILFTEATRILFFNTPKKMTDYAKKRGWVLGPNNYYSFASQQQKPEDTTIPSTELAKQVIEYARQLEMIV
Specifications

Specifications

proteasome (prosome, macropain) 26S subunit, non-ATPase, 8
Unconjugated
No additive
NM_002812
HIP6|HYPF|MGC1660|Nin1p|Rpn12|S14|p31
Mouse
50 uL
Cell Cycle
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human PSMD8 protein.
PSMD8
Serum
PSMD8
PSMD8 (NP_002803, 1 a.a. ~ 257 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5714
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit