Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PSMD8, Polyclonal, Invitrogen™

Catalog No. PIPA128267
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA128267 50 μg
1 options

Catalog No. PIPA128267

Supplier: Thermo Scientific PA128267

Chicken Polyclonal Antibody

The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in a non-lysosomal pathway. An essential function of a modified proteasome, the immunoproteasome, is the processing of class I MHC peptides. This gene encodes a non-ATPase subunit of the 19S regulator. A pseudogene has been identified on chromosome 1.

PA1-28267 detects PSMD8 from human, rat, mouse samples.PA1-28267 has been successfully used in Western blot applications.The PA1-28267 immunogen is: Amino acids 1-257 of PSMD8.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PSMD8
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Gene PSMD8
Gene Accession No. P48556, Q9CX56
Gene Alias HIP6, HYPF, MGC1660, Nin1p, Rpn12, S14, p31
Gene Symbols PSMD8
Host Species Chicken
Immunogen Amino acids 1-257 of PSMD8.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5714
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.