Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PSMG2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA526702
Description
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.Specifications
PSMG2 | |
Polyclonal | |
Liquid | |
PSMG2 | |
CLAST3, HCCA3, HsT1707, MDS003, PAC2, TNFSF5IP1, CD40 ligand-activated specific transcript 3, HDCMC29P, HSPC260, PAC-2, hepatocellular carcinoma susceptibility protein, hepatocellular carcinoma-susceptibility protein 3, likely ortholog of mouse CD40 ligand-activated specific transcript 3 (Clast3), proteasome a | |
PSMG2 | |
KLH conjugated synthetic peptide between 217-246 amino acids from the C-terminal region of human PSMG2 | |
Polyclonal | |
Antigen affinity chromatography | |
RUO | |
Human |
Western Blot | |
Unconjugated | |
PBS with 0.09% sodium azide | |
Q969U7 | |
56984 | |
Rabbit | |
IgG | |
Primary | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Antibody |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title