Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A7 Mouse anti-Human, Clone: 47C1068, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA191555

Catalog No. MA191555


Add to cart

Description

Description

MA1-91555 detects Psoriasin from human samples. MA1-91555 has been successfully used in immunohistochemistry & Western blot applications, where a band of ∽11 kDa is detected. Confluent MCF10A cells can be used as a positive control for this antibody. The MA1-91555 immunogen is recombit human full length protein. Store at 4°C short term. For long term storage, aliquot and store at -20°C or below, avoiding freeze-thaw cycles.

The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. S100 genes include at least 13 members which are located as a cluster on chromosome 1q21. This protein differs from the other S100 proteins of known structure in its lack of calcium binding ability in one EF-hand at the N-terminus. This protein is markedly over-expressed in the skin lesions of psoriatic patients, but is excluded as a candidate gene for familial psoriasis susceptibility. The exact function of this protein is not known.
Specifications

Specifications

S100A7
Monoclonal
1.0 mg/mL
PBS with 0.05% sodium azide
P31151
PSOR1, S100A7c
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6278
Immunohistochemistry (Paraffin), Western Blot
47C1068
Unconjugated
S100A7
Liquid
S100A7
Recombit full length protein (Human).
Protein G
RUO
Antibody
Monoclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit