Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PSPC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535958
Description
Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (94%), Rat (94%), Rhesus Monkey (97%), Bovine (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.
Specifications
PSPC1 | |
Polyclonal | |
Unconjugated | |
PSPC1 | |
5730470C09Rik; AI327109; AI449052; mPSP1; Paraspeckle component 1; paraspeckle protein 1; PSP1; Pspc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
305910, 55269, 66645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q4KLH4, Q8R326, Q8WXF1 | |
PSPC1 | |
Recombinant fragment corresponding to a region within amino acids 1 and 176 of PSPC1. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction