Learn More
MilliporeSigma™ anti-PTEN, Clone: 6H2.1, ™ (Upstate™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ 04035/DEL
Catalog No. 50-171-125
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
The protein and lipid phosphatase PTEN (phosphatase and tensin homolog deleted on chromosome ten) has been suggested to play a role in tumorgenesis. PTEN is a tumor suppressor whose loss permits constitutive signaling through the PI3K pathway and is suffi
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Specifications
Specifications
PTEN
1mg/mL
Semi-purified
C-terminal 100 amino acids of PTEN
100μg
Cell Signaling
Monoclonal
6H2.1
Unconjugated
Murine
IgG
RUO
Primary
Human, Murine, Rat