Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTF1A Mouse, Alexa Fluor 647, Clone: S25-763, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564750

Catalog No. BDB564750


Add to cart

Description

Description

The S25-763 monoclonal antibody specifically binds PTF1A (Pancreas specific transcription factor 1a). PTF1A is a transcription factor that contains a basic helix-loop-helix motif domain and, in combination with p75 and p64, forms the pancreas transcription factor complex. PTF1A plays a roles in pancreatic and cerebellar development. In early pancreatic development, pancreatic bud cells can either continue to pancreatic organogenesis or regress to a duodenal fate, and PTF1A plays a role in this differentiation. In late pancreatic development, the expression of PTF1A is restricted to acinar cells where it is involved in the maintenance of exocrine-specific gene expression. PTF1A expression is often evaluated to monitor pancreatic lineage differentiation from pluripotent stem cells. In addition, PTF1A is expressed during retinal development, and mutations in PTF1A cause cerebellar deficiencies.

This mouse anti-human PTF1A monoclonal antibody, clone S25-763, has been demonstrated to cross-react with rat and mouse pancreatic acinar cells.

Specifications

Specifications

PTF1A
S25-763
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Murine
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
PTF1α; Ptf1a; Ptf1α; bHLHa29; PTF1-p48
Affinity Purified
Human PTF1A Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit