Learn More
PTH/PTHrP Receptor Mouse anti-Human, Mouse, Pig, Clone: 3D1.1, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05517
Catalog No. 05-517-MI
Add to cart
Description
Description
Anti-PTH/PTHrP Receptor Antibody, clone 3D1.1 is an antibody against PTH/PTHrP Receptor for use in immunohistochemistry and western blotting.
Specifications
Specifications
PTH/PTHrP Receptor
3D1.1
Protein G Purified immunoglobulin in Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
200μg
Primary
Human, Murine, Porcine
Immunohistochemistry, Western Blot
Unconjugated
NM_000315
PTHLH, PTHRP, osteostatin, PTHrP, MGC14611, PTHR, 107-139)., PLP, HHM, PTH-rP
Synthetic peptide corresponding to amino acids 146-169 of the human PTH/PTHrP (parathyroid hormone/parathyroid hormone related protein) receptor (RRCDRNGSWELVPGHNRTWANYSE).
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal