Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

twinfilin, actin-binding protein, homolog 1 (Drosophila), Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human PTK9 protein.

Manufacturer:  Abnova Corporation H00005756B01

Catalog No. 89-006-088


Add to cart

Description

Description

This gene encodes twinfilin, an actin monomer-binding protein conserved from yeast to mammals. Studies of the mouse counterpart suggest that this protein may be an actin monomer-binding protein, and its localization to cortical G-actin-rich structures may be regulated by the small GTPase RAC1. [provided by RefSeq

Sequence: MTRRGRRRSSHFLGPPAGAAGCTQRRSRELAAAAMSHQTGIQASEDVKEIFARARNGKYRLLKISIENEQLVIGSYSQPSDSWDKDYDSFVLPLLEDKQPCYILFRLDSQNAQGYEWIFIAWSPDHSHVRQKMLYAATRATLKKEFGGGHIKDEVFGTVKEDVSLHGYKKYLLSQSSPAPLTAAEEELRQIKINEVQTDVGVDTKHQTLQGVAFPISREAFQALEKLNNRQLNYVQLEIDIKNEIIILANTTNTELKDLPKRIPKDSARYHFFLYKHSHEGDYLESIVFIYSMPGYTCSIRERMLYSSCKSRLLEIVERQLQMDVIRKIEIDNGDELTADFLYEEVHPKQHAHKQSFAKPKGPAGKRGIRRLIRGPAETEATTD
Specifications

Specifications

twinfilin, actin-binding protein, homolog 1 (Drosophila)
Unconjugated
No additive
NM_002822.3
A6|MGC23788|MGC41876|PTK9
Mouse
50 uL
Kinases
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human PTK9 protein.
TWF1
Serum
TWF1
PTK9 (NP_002813.2, 1 a.a. ~ 384 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
5756
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit