Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTOP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132165
Description
PTOP Polyclonal specifically detects PTOP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PTOP | |
Polyclonal | |
Unconjugated | |
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
CDK15 | |
Recombinant protein encompassing a sequence within the center region of human PFTK2. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
65061 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.57mg/mL | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
ALS2CR7, amyotrophic lateral sclerosis 2 (juvenile) chromosome region, candidate 7, Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 7 protein, Cell division protein kinase 15, cyclin-dependent kinase 15, EC 2.7.11, EC 2.7.11.22, PFTAIRE protein kinase 2, PFTK2, Serine/threonine-protein kinase ALS2CR7, Serine/threonine-protein kinase PFTAIRE-2 | |
Rabbit | |
49 kDa | |
0.1 mL | |
Primary | |
Tested in human. Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction