Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTOV1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524243

Catalog No. PIPA524243

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine based on sequence homology.

May activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
Specifications

Specifications

PTOV1
Polyclonal
PBS with 0.09% sodium azide
Q86YD1, Q91VU8
ACID2, PTOV-1, activator interaction domain-containing protein 2, prostate tumor overexpressed gene 1, prostate tumor-overexpressed gene 1 protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
53635, 84113
Immunohistochemistry, Western Blot
Unconjugated
PTOV1
Liquid
PTOV1
KLH conjugated synthetic peptide between 25-52 amino acids from the N-terminal region of human PTOV1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit