Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEG2 Mouse anti-Human, Mouse, Rat, Clone: 291835, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523997

Catalog No. PIMA523997


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

MEG2 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. This PTP contains an N-terminal domain that shares a significant similarity with yeast SEC14, which is a protein that has phosphatidylinositol transfer activity and is required for protein secretion through the Golgi complex in yeast. This PTP was found to be activated by polyphosphoinositide, and is thought to be involved in signaling events regulating phagocytosis.
Specifications

Specifications

MEG2
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P43378, O35239, Q641Z2
Tyrosine-protein phosphatase, non-receptor type 9, Protein-tyrosine phosphatase MEG2, PTPase-MEG2
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
266611, 56294, 5780
Immunocytochemistry, Immunofluorescence, Western Blot
291835
Unconjugated
PTPN9
Lyophilized
PTPN9
E. coli-derived recombit human PTP-MEG2 Glu2-Gln593
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit