Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPα Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07472
Description
Anti-PTPα Antibody detects level of PTPα and has been published and validated for use in western blotting.
Specifications
PTPα | |
Polyclonal | |
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30% | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot | |
Unconjugated | |
PTPRA; RPTPA; HLPR; PTPRL2; R-PTP-alpha; PTPA; HPTPA; HEPTP; HPTPalpha; LRP; PTPase-alpha | |
N-terminal GST-tagged fusion protein corresponding to residues 505-793 of human PTP alpha (complete D2 catalytic domain). | |
100 μL | |
NM_178001 | |
−20°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction