Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-PTP sigma/PTPRS, Polyclonal, Novus Biologicals™

Supplier: Novus Biologicals 37160002
Specifications
PTP sigma/PTPRS | |
Polyclonal | |
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000 | |
Q13332 | |
PTPRS | |
This antibody is specific for the Middle Region of the target protein. | |
0.05 mg | |
Primary | |
This product is specific for Human PTPRS. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
Unconjugated | |
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative | |
EC 3.1.3.48, protein tyrosine phosphatase PTPsigma, protein tyrosine phosphatase, receptor type, S, protein tyrosine phosphatase, receptor type, sigma, PTPSIGMA, receptor-type tyrosine-protein phosphatase S, Receptor-type tyrosine-protein phosphatase sigma, R-PTP-S, R-PTP-sigma | |
Rabbit | |
Affinity Purified | |
RUO | |
5802 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction