Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

PTP1B Mouse, Unlabeled, Clone: 15, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610140

 View more versions of this product

Catalog No. BDB610140



Description

Description

PTP1B is a member of the class of enzymes termed protein tyrosine phosphatases(PTPs). This 50 kDa protein contains a conserved phosphatases domain at residues 30-278 and is localized to the cytoplasmic face of the endoplasmic reticulum by its C-terminal 35 residues. Its interactions with other proteins are mediated by proline-rich regions and SH2 compatible sequences. PTP1B is thought to act as a negative regulator in insulin signaling. Insulin stimulation induces its association with the insulin receptor and tyrosine phosphorylation. Its overexpression inhibits proximal and distal insulin signaling events, possibly due to receptor dephosphorylation. Microinjection of PTP1B into Xenopus oocytes blocks insulin-induced maturation. Additional studies demonstrate that PTP1B undergoes cell cycle-dependent and/or stress-induced serine phosphorylation. This suggests further regulatory roles that are not yet defined.

Specifications

Specifications

PTP1B
15
Unconjugated
Affinity Purified
Aqueous buffered solution containing BSA
150μg
Cell Biology
Primary
Canine, Chicken, Frog, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.