Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PRL3, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA131531

Catalog No. PIPA131531

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Protein tyrosine phosphatase which stimulates progression from G1 into S phase during mitosis. Enhances cell proliferation. cell motility and invasive activity. and promotes cancer metastasis. May be involved in the progression of cardiac hypertrophy by i

PA1-31531 detects PTP4A3 from human samples.PA1-31531 has been successfully used in Western blot applications.The PA1-31531 immunogen is: Peptide with sequence NRPAPVEVSYKHMR, from N Terminus from the second Met of the protein sequence according to NP_009010; NP_116000
Specifications

Specifications

PTP4A3
Polyclonal
Unconjugated
PTP4A3
Purified
PTP4A3
Peptide with sequence NRPAPVEVSYKHMR, from N Terminus from the second Met of the protein sequence according to NP_009010; NP_116000
IgG fraction
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
0.5mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
O75365, Q9D658
PRL-3, PRL-R, PRL3
Goat
IgG
50μg
Avoid Freeze/Thaw Cycles
Primary
11156
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit