Learn More
PTPA Mouse anti-Human, Mouse, Clone: 5G3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05941
Catalog No. 05-941-MI
Add to cart
Description
Description
Detect PTPA using this Anti-PTPA Antibody, clone 5G3 validated for use in western blotting.
Specifications
Specifications
PTPA
5G3
NM_178001
PPP2R4, PR53, KIAA0044, PTPA, MGC2184, PP2A
Full length mouse phosphotyrosyl phosphatase activator protein (PTPA)
−20°C for 24 months.
Monoclonal
Western Blot
Unconjugated
Supernatant
Murine
100μL
Primary
Human, Murine