Learn More
PTPLB Rabbit anti-Hamster, Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524897
Catalog No. PIPA524897
Add to cart
Description
Description
This antibody is predicted to react with bovine based on sequence homology.
PTPLB is probable anti-phosphatase.
Specifications
Specifications
PTPLB
Polyclonal
Unconjugated
HACD2
Liquid
Hacd2
KLH conjugated synthetic peptide between 226-254 amino acids from the C-terminal region of human PTPLB
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Hamster, Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6Y1H2, Q9D3B1
HACD2, 3-hydroxyacyl-CoA dehydratase 2, protein-tyrosine phosphatase-like member B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
100765668, 201562, 70757