Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PTPN13/PTPL1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$216.50 - $361.50

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A recombinant protein corresponding to amino acids 1279 to 1883 of human FAP-1 protein was used as immunogen; GenBank no. NP_542414.1.
Antigen PTPN13/PTPL1
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056139T
View Documents
Novus Biologicals
NB10056139SS
0.025 ml
Each for $216.50
Add to cart
 
NB10056139
View Documents
Novus Biologicals
NB10056139
0.05 ml
Each for $361.50
Add to cart
 
Description

Description

PTPN13/PTPL1 Polyclonal antibody specifically detects PTPN13/PTPL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Rabbit
A recombinant protein corresponding to amino acids 1279 to 1883 of human FAP-1 protein was used as immunogen; GenBank no. NP_542414.1.
Polyclonal
Primary
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Q12923
Neat whole antisera with 0.05% Sodium Azide
IgG
PTPN13/PTPL1
Antisera
RUO
FAP-1 typically migrates at ∼270 kDa on western blots. Thus for SDS-PAGE and western blot, the researcher should use techniques optimized for the transfer of proteins with high molecular weights. The actual molecular weight of FAP-1 observed depends on the isoform expression.
5783
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit