Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN22, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127642

Catalog No. PIPA127642

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This protein is a protein tyrosine phosphatase which is expressed primarily in lymphoid tissues. This enzyme associates with the molecular adapter protein CBL and may be involved in regulating CBL function in the T-cell receptor signaling pathway. Alterna

PA1-27642 detects PTPN22 from human samples.PA1-27642 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Protein tyrosine phosphatase. non-receptor type 22.
Specifications

Specifications

PTPN22
Polyclonal
PBS with no preservative
Q9Y2R2
LYP, LYP1, LYP2, PEP, PTPN8
Chicken
IgY
50μg
Store at -20°C
Primary
26191
Western Blot
Unconjugated
PTPN22
Purified
PTPN22
Synthetic peptide corresponding to human Protein tyrosine phosphatase. non-receptor type 22.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit