Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTPN7 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514559

Catalog No. PIPA514559


Add to cart

Description

Description

PTPN7 is a member of the protein tyrosine phosphatase (PTP) family. PTPs are known to be signaling molecules that regulate a variety of cellular processes including cell growth, differentiation, mitotic cycle, and oncogenic transformation. PTPN7 is preferentially expressed in a variety of hematopoietic cells, and is an early response gene in lymphokine stimulated cells. The noncatalytic N-terminus of this PTP can interact with MAP kinases and suppress the MAP kinase activities. This PTP has been shown to be involved in the regulation of T cell antigen receptor (TCR) signaling, which is thought to function through dephosphorylating the molecules related to MAP kinase pathway.
Specifications

Specifications

PTPN7
Polyclonal
PBS with 0.09% sodium azide
P35236
Tyrosine-protein phosphatase non-receptor type 7; Protein-tyrosine phosphatase LC-PTP; Hematopoietic protein-tyrosine phosphatase; LPTP; HEPTP; PTPNI; BPTP-4; LC-PTP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5778
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PTPN7
Liquid
PTPN7
KLH conjugated synthetic peptide between 22-51 amino acids from human PTPN7
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit