Learn More
CD148, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127625
Description
CD148 is a highly glycosylated transmembrane protein of up to 250 kDa, containing eight fibronectin type III domains in the extracellular region and one tyrosine phosphatase domain in the cytoplasmic region. Expression is found broadly on fibroblasts, epithelial cells, lymphocytes (B, T and NK cells), and myeloid cells (monocytes and granulocytes). CD148 can dephosphorylate proteins such as LAT and PLC-gamma, and is involved in the downregulation of T cell activation. Conversely, CD148 expression can be upregulated following T cell activation.
PA1-27625 detects PTPRJ from human samples.PA1-27625 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Receptor-type tyrosine-protein phosphatase eta.
Specifications
PTPRJ/DEP1 | |
Polyclonal | |
PTPRJ | |
CD148, DEP1, HPTPeta, R-PTP-ETA, SCC1 | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
5795 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
Q12913 | |
PTPRJ | |
Synthetic peptide corresponding to human Receptor-type tyrosine-protein phosphatase eta. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.