Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PTPRZ, Polyclonal, Invitrogen™
Chicken Polyclonal Antibody
Supplier: Thermo Scientific PA127632
Description
PA1-27632 detects PTPRZ2 from human samples.PA1-27632 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The immunogen is a synthetic peptide corresponding to human Tyrosine phosphatase zeta polypeptide 2 HTPZP2.Specifications
PTPRZ2 | |
Polyclonal | |
PTPRZ1 | |
HTPZP2, PTPRZ | |
Chicken | |
Antigen affinity chromatography | |
RUO | |
5803 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Western Blot | |
Unconjugated | |
P23471 | |
PTPRZ1 | |
Synthetic peptide corresponding to human Tyrosine phosphatase zeta polypeptide 2 HTPZP2. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgY |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction