Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PTRF Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514319

Catalog No. PIPA514319

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Termination of transcription by RNA polymerase I involves pausing of transcription by TTF1, and the dissociation of the transcription complex, releasing pre-rRNA and RNA polymerase I from the template. PTRF is required for dissociation of the ternary transcription complex.
Specifications

Specifications

PTRF
Polyclonal
Unconjugated
CAVIN1
Liquid
CAVIN1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human PTRF
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 0.09% sodium azide
Q6NZI2
Polymerase I and transcript release factor; PTRF; FKSG13
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
284119
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit