Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-PUMA, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$119.60 - $314.00

Specifications

Host Species Rabbit
Isotype IgG
Immunogen Synthetic peptide made to an N-terminal portion of human PUMA (within residues 1-100). [NCBI Sequence NP_001120712]
Antigen PUMA
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11081760T
View Documents
Novus Biologicals
NB11081760SS
0.025 ml
Each for $119.60
Add to cart
 
NB11081760
View Documents
Novus Biologicals
NB11081760
0.1 ml
Each for $314.00
Add to cart
 
Description

Description

PUMA Polyclonal antibody specifically detects PUMA in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Rabbit
Synthetic peptide made to an N-terminal portion of human PUMA (within residues 1-100). [NCBI Sequence NP_001120712]
Polyclonal
Primary
Western Blot, Immunocytochemistry, Immunofluorescence
NP_001120712
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide
IgG
PUMA
Affinity Purified
RUO
This is specific for isoform 1 of PUMA.
27113
1 mg/mL
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit