Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PUMA alpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA14277
Description
PA1-4277 detects alpha- and beta-PUMA from human samples. PA1-4277 has been successfully used in Western blot procedures. By Western blot this antibody recognizes both forms of the PUMA protein found at ∽20 kDa and ∽36 kDa in transfected U20S cells. PA1-4277 immunizing peptide corresponds to amino acid residues E(101) Q H L E S P V P S A P G A L A G(117) of the human PUMA-a form of the PUMA protein.
PUMA alpha Polyclonal antibody specifically detects PUMA alpha in Human samples. It is validated for Western Blot.Specifications
PUMA alpha | |
Polyclonal | |
Unconjugated | |
BBC3 | |
BBC3; BCL2 binding component 3; Bcl-2 binding component 3; bcl-2-binding component 3; Bcl-2-binding component 3, isoforms 1/2; Bcl-2-binding component 3, isoforms 3/4; BH3-only; FLJ42994; JFY1; JFY-1; p53 up-regulated modulator of apoptosis; PUMA; PUMA/JFY1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27113 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q96PG8, Q9BXH1 | |
BBC3 | |
Synthetic Peptide: E(101) Q H L E S P V P S A P G A L A G(117) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction