Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PFAS Rabbit anti-Human, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer: Invitrogen PA524685
Description
This antibody is predicted to react with mouse based on sequence homology.
Specifications
PFAS | |
Polyclonal | |
PBS with 0.09% sodium azide | |
O15067 | |
FGAMS, FGARAT, PURL, FGAM synthase, FGAR amidotransferase, formylglycinamide ribotide amidotransferase, formylglycinamide ribotide synthetase | |
Rabbit | |
IgG | |
400 μL | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
5198 |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
PFAS | |
Liquid | |
PFAS | |
KLH conjugated synthetic peptide between 184-213 amino acids from the N-terminal region of human PUR4 | |
Antigen affinity chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title