Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

P2Y2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA146150

Catalog No. PIPA146150

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Suggested positive control: human brain lysate and rat dorsal root ganglion, antigen standard for P2RY2 (transient overexpression lysate).

P2Y2 belongs to the family of G-protein coupled receptors. This family has several receptor subtypes with different pharmacological selectivity, which overlaps in some cases, for various adenosine and uridine nucleotides. This receptor is responsive to both adenosine and uridine nucleotides. It may participate in control of the cell cycle of endometrial carcinoma cells. P2Y2 is also a morphogen receptor that potentiates neurotrophin signaling in neuronal development and regeneration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

P2Y2
Polyclonal
Unconjugated
P2RY2
P2RY2
Rabbit
Antigen affinity chromatography
RUO
18442, 29597, 5029
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 0.05% sodium azide
P41231, P35383, P41232
P2RY2
Synthetic peptide comprising residues 351-366 of the human P2Y2 protein.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit