Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PUS3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524321

Catalog No. PIPA524321


Add to cart

Description

Description

Formation of pseudouridine at position 39 in the anticodon stem and loop of transfer RNAs.
Specifications

Specifications

PUS3
Polyclonal
PBS with 0.09% sodium azide
Q9BZE2
2610020J05Rik, tRNA pseudouridine synthase 3, tRNA pseudouridine(38/39) synthase, tRNA pseudouridylate synthase 3, tRNA-uridine isomerase 3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
83480
Immunohistochemistry, Western Blot
Unconjugated
PUS3
Liquid
PUS3
KLH conjugated synthetic peptide between 35-64 amino acids from the N-terminal region of human PUS3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit