Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PXMP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526188

Catalog No. PIPA526188

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PXMP4 belongs to the peroxisomal membrane protein PXMP2/4 family.
Specifications

Specifications

PXMP4
Polyclonal
Unconjugated
PXMP4
Liquid
PXMP4
KLH conjugated synthetic peptide between 128-155 amino acids from the central region of human PXMP4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
Q9Y6I8, Q9JJW0
PMP24, 24 kDa peroxisomal intrinsic membrane protein, peroxisomal membrane protein 4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
11264, 59038
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit