Learn More
Pyk2/CAKΒ Mouse anti-Human, Rat, Clone: 74, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05488
Catalog No. 05-488-MI
Add to cart
Description
Description
Anti-Pyk2/CAKβ Antibody, clone 74 is an antibody against Pyk2/CAKβ for use in immunoprecipitation and western blotting.
Specifications
Specifications
Pyk2/CAKβ
74
250μL of mouse ascites containing 0.05% sodium azide. Frozen at −20°C.
Ascites
Mouse
IgG1
250μL
Primary
Human, Rat
Immunoprecipitation, Western Blot
Unconjugated
NM_173174
PTK2B, PYK2, RAFTK, FADK2, PKB, FAK2, CAKB, PTK, CADTK
GST fusion protein corresponding to residues 5-416 of rat Pyk2
Thiophilic adsorption chromatography
−20°C for 24 months.
Monoclonal